APO455836229 Apollo Scientific Ltd

4,5-Difluoro-2-nitrotoluene

  • MDL number MFCD04973772
  • Cas Number : 127371-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5-Difluoro-2-methylnitrobenzene, 1,2-Difluoro-4-methyl-5-nitrobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-2-nitrotoluene

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-2-nitrotoluene
pictograms GHS06
hstatements H301+H311+H331;H319
Availability details for
Quantity
  • Availability
  • Add to Cart