APO455836230 Apollo Scientific Ltd

1-[4-(Trifluoromethyl)phenyl]propane-1,2-dione

  • MDL number MFCD01941310
  • Cas Number : 10557-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(2-Oxopropanoyl)benzotrifluoride, 2-Oxo-4'-(trifluoromethyl)propiophenone
Purity 95
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 70/0.2mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5427

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5427
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart