APO455836248 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD00040980
  • Cas Number : 115029-22-6
  • Molecular Weight : 208.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Synonym 3-Carboxy-2-fluorobenzotrifluoride
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart