APO455836257 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD00061280
  • Cas Number : 4088-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-Tetrafluoro-m-tolunitrile
Purity 97
Density 1.38
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 186
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzonitrile-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylbenzonitrile-97
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart