APO455836261 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD00061284
  • Cas Number : 67515-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-69
Synonym 5-Cyano-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-Tetrafluoro-m-tolunitrile
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 193-195
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzonitrile-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzonitrile-97
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart