APO455836278 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD00061166
  • Cas Number : 207981-45-1
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24-26
Purity 97
Density 1.39
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 211
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart