APO455836282 Apollo Scientific Ltd

4-Fluoro-2-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD00061170
  • Cas Number : 220227-29-2
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 55-57
Iupac Name [4-fluoro-2-(trifluoromethyl)phenyl]methanol
Inchi Code 1S/C8H6F4O/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3,13H,4H2
Inchi Key QCDWCMMCVRWROH-UHFFFAOYSA-N

Description

Synonym 5-Fluoro-2-(hydroxymethyl)benzotrifluoride
Purity 97%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 55-57
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-trifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-trifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart