APO455836289 Apollo Scientific Ltd

3-Fluoro-5-(trifluoromethyl)benzyl bromide

  • MDL number MFCD00061175
  • Cas Number : 239087-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 43/1mm
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylbenzyl-bromide-98

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylbenzyl-bromide-98
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart