APO455836290 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethyl)benzyl bromide

  • MDL number MFCD00061177
  • Cas Number : 184970-26-1
  • Molecular Weight : 257.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene
Inchi Code 1S/C8H5BrF4/c9-4-5-1-2-7(10)6(3-5)8(11,12)13/h1-3H,4H2
Inchi Key JVQDTRQMKZMBAU-UHFFFAOYSA-N

Description

Synonym 5-(Bromomethyl)-2-fluorobenzotrifluoride, 4-(Bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart