APO455836293 Apollo Scientific Ltd

2-Fluoro-5-hydroxybenzotrifluoride

  • MDL number MFCD00043875
  • Cas Number : 61721-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 17
Synonym 4-Fluoro-3-(trifluoromethyl)phenol, alpha,alpha,alpha,6-Tetrafluoro-m-cresol
Purity 98
Flash Point 79
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 84-86/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2219

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=2219
pictograms GHS05;GHS07;GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart