APO455836295 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD00061187
  • Cas Number : 220227-66-7
  • Molecular Weight : 222.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-109
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylphenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethylphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart