APO455836301 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD00061183
  • Cas Number : 220239-65-6
  • Molecular Weight : 203.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 93/10mm
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylphenylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylphenylacetonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart