APO455836302 Apollo Scientific Ltd

2'-Fluoro-3'-(trifluoromethyl)propiophenone

  • MDL number MFCD00061192
  • Cas Number : 207986-23-0
  • Molecular Weight : 220.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 127-129
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylpropiophenone

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylpropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart