APO455836303 Apollo Scientific Ltd

2'-Fluoro-4'-(trifluoromethyl)propiophenone

  • MDL number MFCD00061259
  • Cas Number : 208173-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2-Fluoro-4-(trifluoromethyl)phenyl]propan-1-one, 1-[2-Fluoro-4-(trifluoromethyl)phenyl]-1-oxopropane
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2238

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2238
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart