APO455836311 Apollo Scientific Ltd

1,2-Dimethyl-3-fluorobenzene

  • MDL number MFCD00000323
  • Cas Number : 443-82-3
  • Molecular Weight : 124.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoro-o-xylene
Purity 98
Density 0.992
Flash Point 36
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 147-152
Product Link https://store.apolloscientific.co.uk/product/12-dimethyl-3-fluorobenzene

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/12-dimethyl-3-fluorobenzene
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart