APO455836313 Apollo Scientific Ltd

1,3-Dimethyl-5-fluorobenzene

  • MDL number MFCD00052366
  • Cas Number : 461-97-2
  • Molecular Weight : 124.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Fluoro-m-xylene
Purity 98
Flash Point 35
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 144-145
Product Link https://store.apolloscientific.co.uk/product/13-dimethyl-5-fluorobenzene

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-dimethyl-5-fluorobenzene
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart