APO455836315 Apollo Scientific Ltd

2-[(4-Ethoxyphenyl)amino]-4-(4-fluoro-3-methylphenyl)-1,3-thiazole

  • MDL number MFCD00129327
  • Cas Number : 1092346-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(4-ethoxyphenyl)-4-(4-fluoro-3-methylphenyl)-1,3-thiazol-2-amine
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1567

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1567
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart