APO455836324 Apollo Scientific Ltd

3-Fluoro-4-nitrobenzoic acid

  • MDL number MFCD01862092
  • Cas Number : 403-21-4
  • Molecular Weight : 185.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Synonym 4-Carboxy-2-fluoronitrobenzene
Purity 97
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart