APO455836325 Apollo Scientific Ltd

1H,1H-Heptafluoro-1-iodobutane

  • MDL number MFCD00013719
  • Cas Number : 374-98-1
  • Molecular Weight : 309.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1,1,2,2,3,3-Heptafluoro-4-iodobutane, (Perfluoropropyl)methyl iodide
Purity 97
Density 2.05
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91-92
Product Link https://store.apolloscientific.co.uk/product/1h1h-heptafluoro-1-iodobutane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h-heptafluoro-1-iodobutane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart