APO455836330 Apollo Scientific Ltd

4-Amino-3-fluorobenzyl alcohol

  • MDL number MFCD03094241
  • Cas Number : 146019-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-4-(hydroxymethyl)aniline, (4-Amino-3-fluorophenyl)methanol
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart