APO455836339 Apollo Scientific Ltd

2-Fluoro-5-nitroanisole

  • MDL number MFCD07782081
  • Cas Number : 454-16-0
  • Molecular Weight : 171.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 66-71
Iupac Name 1-fluoro-2-methoxy-4-nitrobenzene
Inchi Code 1S/C7H6FNO3/c1-12-7-4-5(9(10)11)2-3-6(7)8/h2-4H,1H3
Inchi Key AZNKKZHZGDZSIF-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 4-Fluoro-3-methoxynitrobenzene, 1-Fluoro-2-methoxy-4-nitrobenzene, 2-Fluoro-5-nitrophenyl methyl ether
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 66-71
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-nitroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart