APO455836340 Apollo Scientific Ltd

2'-Fluoro-4'-hydroxy-5'-methoxyacetophenone

  • MDL number MFCD11101445
  • Cas Number : 1065076-49-4
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 113-114
Iupac Name 1-(2-fluoro-4-hydroxy-5-methoxyphenyl)ethanone
Inchi Code 1S/C9H9FO3/c1-5(11)6-3-9(13-2)8(12)4-7(6)10/h3-4,12H,1-2H3
Inchi Key JTOSXEWSXIZJGX-UHFFFAOYSA-N

Description

Synonym 1-(2-Fluoro-4-hydroxy-5-methoxyphenyl)ethan-1-one, 4-Acetyl-5-fluoro-2-methoxyphenol, 5-Acetyl-4-fluoro-2-hydroxyanisole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 113-114
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxy-5-methoxyacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxy-5-methoxyacetophenone
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart