APO455836346 Apollo Scientific Ltd

alpha-Methyl-4-(trifluoromethyl)benzylamine

  • MDL number MFCD04038878
  • Cas Number : 15996-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[4-(Trifluoromethyl)phenyl]ethylamine, 4-(1-Aminoethyl)benzotrifluoride
Purity 98
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8395

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=8395
Availability details for
Quantity
  • Availability
  • Add to Cart