APO455836351 Apollo Scientific Ltd

Methyl 3-(trifluoromethyl)benzoate

  • MDL number MFCD00043543
  • Cas Number : 2557-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Methoxycarbonyl)benzotrifluoride, Methyl alpha,alpha,alpha-trifluoro-m-toluate
Purity 99
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11513

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=11513
Availability details for
Quantity
  • Availability
  • Add to Cart