APO455836366 Apollo Scientific Ltd

Heptafluoro-1-(1-iodo-1,2,2,2-tetrafluoroethoxy)propane

  • MDL number MFCD00155845
  • Cas Number : 107432-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.9859
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 84-85
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3582

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3582
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart