APO455836370 Apollo Scientific Ltd

3-Fluorophenethyl bromide

  • MDL number MFCD03095397
  • Cas Number : 25017-13-4
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromoethyl)-3-fluorobenzene
density 1.45
Inchi Code 1S/C8H8BrF/c9-5-4-7-2-1-3-8(10)6-7/h1-3,6H,4-5H2
Inchi Key GLVSPVSJMYQIPJ-UHFFFAOYSA-N

Description

Synonym 1-(2-Bromoethyl)-3-fluorobenzene, 1-Bromo-2-(3-fluorophenyl)ethane
Purity 95%
Density 1.45
Flash Point 94/10mm
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluorophenethyl-bromide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluorophenethyl-bromide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart