APO455836372 Apollo Scientific Ltd

1H,1H,9H-Hexadecafluorononan-1-ol

  • MDL number MFCD00039629
  • Cas Number : 376-18-1
  • Molecular Weight : 432.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-60
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluorononan-1-ol
Purity 90
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 155-158/200mm
Product Link https://store.apolloscientific.co.uk/product/1h1h9h-hexadecafluorononan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h9h-hexadecafluorononan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart