APO455836373 Apollo Scientific Ltd

1H,1H,9H-Perfluorononyl methacrylate

  • MDL number MFCD00078427
  • Cas Number : 1841-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H,9H-Hexadecafluorononyl methacrylate
Purity 97
Density 1.618
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 234
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4532

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4532
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart