APO455836386 Apollo Scientific Ltd

4-(Trifluoromethyl)-DL-phenylalanine

  • MDL number MFCD00070813
  • Cas Number : 14091-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-202
Synonym 2-Amino-3-[4-(trifluoromethyl)phenyl]propanoic acid, 4-(2-Amino-2-carboxyethyl)benzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethyl-dl-phenylalanine-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethyl-dl-phenylalanine-97
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart