APO455836391 Apollo Scientific Ltd

2,2,4,4-Tetrafluoro-1,3-dithietane

  • MDL number MFCD00053538
  • Cas Number : 1717-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.582
Flash Point 35
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 47-50
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4801

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4801
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart