APO455836392 Apollo Scientific Ltd

2-Fluorophenacylamine hydrochloride

  • MDL number MFCD08532458
  • Cas Number : 93102-96-6
  • Molecular Weight : 189.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-203
Synonym 2-Amino-2'-fluoroacetophenone hydrochloride, 2-Amino-1-(2-fluorophenyl)ethan-1-one hydrochloride
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluorophenacylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluorophenacylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart