APO455836394 Apollo Scientific Ltd

5-Fluoro-2-methylphenol

  • MDL number MFCD00673009
  • Cas Number : 452-85-7
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-hydroxytoluene, 5-Fluoro-o-cresol
Purity 98
Density 1.15
Flash Point 85
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 66/4mm
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-methylphenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-methylphenol
pictograms GHS05;GHS07
hstatements H302+H312;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart