APO455836397 Apollo Scientific Ltd

2-(Trifluoromethoxy)chlorobenzene

  • MDL number MFCD03840206
  • Cas Number : 450-96-4
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxychlorobenzene

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxychlorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart