APO455836404 Apollo Scientific Ltd

2-Methoxy-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD04115975
  • Cas Number : 132927-09-4
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-55
Synonym 4-Formyl-3-methoxybenzotrifluoride, 2-Formyl-5-(trifluoromethyl)anisole, 4-(Trifluoromethyl)-o-anisaldehyde
Purity 99
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-4-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-4-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart