APO455836412 Apollo Scientific Ltd

2-Amino-6-(6-fluoropyridin-2-yl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD00794643
  • Cas Number : 338413-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2312

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2312
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart