APO455836413 Apollo Scientific Ltd

5-Amino-1-(tert-butyl)-3-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD01568265
  • Cas Number : 320422-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2269

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2269
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart