APO455836414 Apollo Scientific Ltd

2-{1-Amino-2-[3-(trifluoromethyl)phenoxy]ethylidene}malononitrile

  • MDL number MFCD00139203
  • Cas Number : 343375-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2332

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2332
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart