APO455836416 Apollo Scientific Ltd

4-Amino-2,5-difluorobenzonitrile

  • MDL number MFCD00190102
  • Cas Number : 112279-61-5
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-94
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-25-difluorobenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-25-difluorobenzonitrile
pictograms GHS06;GHS07
hstatements H301;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart