APO455836418 Apollo Scientific Ltd

O-(2-Chloro-6-fluorobenzyl)hydroxylamine hydrochloride

  • MDL number MFCD01114572
  • Cas Number : 93081-15-3
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-188
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/o-2-chloro-6-fluorobenzylhydroxylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/o-2-chloro-6-fluorobenzylhydroxylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart