APO455836420 Apollo Scientific Ltd

O-(4-Fluorobenzyl)hydroxylamine hydrochloride

  • MDL number MFCD01114581
  • Cas Number : 51572-89-5
  • Molecular Weight : 177.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-235
Synonym 4-[(Aminooxy)methyl]fluorobenzene
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/o-4-fluorobenzylhydroxylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/o-4-fluorobenzylhydroxylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart