APO455836423 Apollo Scientific Ltd

O-[4-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride

  • MDL number MFCD00231881
  • Cas Number : 321574-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 325-328
Synonym 1-[(Aminooxy)methyl]-4-(trifluoromethyl)benzene hydrochloride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2347

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2347
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart