APO455836424 Apollo Scientific Ltd

4-Chloro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD01631468
  • Cas Number : 1737-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-165
Synonym 5-Carboxy-2-chlorobenzotrifluoride, 4-Chloro-alpha,alpha,alpha-trifluoro-m-toluic acid
Purity 97
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart