APO455836425 Apollo Scientific Ltd

4-Chloro-2-fluoro-5-methylphenacyl bromide

  • MDL number MFCD01935937
  • Cas Number : 338982-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Synonym 2-Bromo-4'-chloro-2'-fluoro-5'-methylacetophenone, 2-Bromo-1-(4-chloro-2-fluoro-5-methylphenyl)ethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2414

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2414
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart