APO455836426 Apollo Scientific Ltd

3-(2-Bromoethoxy)benzotrifluoride

  • MDL number MFCD00215501
  • Cas Number : 18800-39-0
  • Molecular Weight : 269.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym beta-Bromo-3-(trifluoromethyl)phenetole,1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 116-118/10mm
Product Link https://store.apolloscientific.co.uk/product/3-2-bromoethoxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-2-bromoethoxybenzotrifluoride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart