APO455836438 Apollo Scientific Ltd

2,3-Dihydro-8-fluoroquinolin-4(1H)-one

  • MDL number MFCD07784308
  • Cas Number : 38470-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-127
Synonym 8-Fluoro-4-oxo-1,2,3,4-tetrahydroquinoline
Purity 97
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10688

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10688
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart