APO455836439 Apollo Scientific Ltd

Fluoromethyl 2H-hexafluoroprop-2-yl ether

  • MDL number MFCD00153189
  • Cas Number : 28523-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Fluoromethoxy)-1,1,1,3,3,3-hexafluoropropane, 1,1,1,4-Tetrafluoro-2-(trifluoromethyl)-3-oxabutane, Sevoflurane
Density 1.505
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 58
Product Link https://store.apolloscientific.co.uk/product/fluoromethyl-2h-hexafluoroprop-2-yl-ether

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/fluoromethyl-2h-hexafluoroprop-2-yl-ether
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart