APO455836453 Apollo Scientific Ltd

2-Fluorophenyl acetate

  • MDL number MFCD00672912
  • Cas Number : 29650-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Acetoxy-2-fluorobenzene
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2252

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2252
pictograms GHS05;GHS07
hstatements H302+H312+H332;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart