APO455836461 Apollo Scientific Ltd

5-(4-Methoxyphenyl)-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD04039275
  • Cas Number : 380905-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-141
Synonym 4-[3-(Trifluoromethyl)-1H-pyrazol-5-yl]anisole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8505

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8505
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart