APO455836475 Apollo Scientific Ltd

4-Amino-3-nitrophenylsulphur pentafluoride

  • MDL number MFCD11501034
  • Cas Number : 197385-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Nitro-4-(pentafluorosulphanyl)aniline, 2-Amino-5-(pentafluorothio)nitrobenzene
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44195

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44195
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart