APO455836477 Apollo Scientific Ltd

2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine

  • MDL number MFCD03424472
  • Cas Number : 111234-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8461

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8461
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart